missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium sulfate, anhydrous, Puratronic™, 99.993% (metals basis)

Manufacturer:  Alfa Aesar™ 1111604

 View more versions of this product

Catalog No. AA1111604



Specifications

Specifications

Calcium sulfate
7778-18-9
CaO4S
MFCD00010912
calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous
[O-]S(=O)(=O)[O-].[Ca+2]
136.134
CHEBI:31346
Powder
Puratronic™
Odorless
White
2g
Anhydrous
(metals basis)
CaSO4
14,1706
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
calcium;sulfate
24497
136.14
99.993%
2.960g/mL
Hygroscopic
1450°C

Chemical Identifiers

7778-18-9
136.134
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
24497
calcium;sulfate
CaO4S
MFCD00010912
calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous
CHEBI:31346
[O-]S(=O)(=O)[O-].[Ca+2]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : WS6920000

TSCA : TSCA

Documents