missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium silicide, tech. Ca ≈30%, may contain up to 5% Fe

Manufacturer:  Alfa Aesar™ 1467636

 View more versions of this product

Catalog No. AA1467636



Specifications

Specifications

Calcium silicide
CaSi2
UN1405
Insoluble in cold water, decomposes in hot water
[Si-].[Si-].[Ca+2]
96.248
96.25
Technical
2.5g/mL
Gray
500g
12013-56-8
MFCD00015977
calciumdisilicid, ? 1-silylcalcio-? 1-silanylidyne
XXCKHUMTXOCLPT-UHFFFAOYSA-N
calcium;silicon(1-)
13848147
Powder
Ca ≈30%, may contain up to 5% Fe
Moisture Sensitive
~1000°C

Chemical Identifiers

12013-56-8
96.248
XXCKHUMTXOCLPT-UHFFFAOYSA-N
13848147
[Si-].[Si-].[Ca+2]
CaSi2
MFCD00015977
calciumdisilicid, ? 1-silylcalcio-? 1-silanylidyne
calcium;silicon(1-)
Safety and Handling

Safety and Handling

GHS H Statement
H261
In contact with water releases flammable gas.

GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM SILICIDE

TSCA : TSCA

SDS Documents