Please login to your online account to display your discounted pricing

Calcium silicate, White powder, Alfa Aesar™

For Use As a Ceramics, Paint Extender, in Chromatography

$51.23 - $274.23

Chemical Identifiers

Chemical Name or Material Calcium silicate
CAS 10101-39-0
Purity Grade Reagent Grade
Physical Form Powder
Molecular Formula CaSiO3
Formula Weight 116.16
MDL Number MFCD00015979
Synonym Calcium metasilicate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA89741A3

  SDS
alfa aesar
89741A3
2kg Each for $274.23

AA8974122

  SDS
alfa aesar
8974122
100g Each for $51.23

AA8974136

  SDS
alfa aesar
8974136
500g Each for $89.80
Description & Specifications

Specifications

Purity Grade Reagent Grade
Physical Form Powder
Formula Weight 116.16
Quantity 2kg
Color White
Density 2.9g/cm3 at 20°C
Chemical Name or Material Calcium silicate
EINECS Number 233-250-6
Molecular Formula CaSiO3
Synonym Calcium metasilicate
CAS 10101-39-0
MDL Number MFCD00015979
Health Hazard 1 H319-H335 : Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Safety and Handling

H319-H335 : Causes serious eye irritation. May cause respiratory irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.