missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium silicate, White powder, Alfa Aesar™

For Use As a Ceramics, Paint Extender, in Chromatography

Manufacturer:  Alfa Aesar 89741A3

 View more versions of this product

Catalog No. AA89741A3



Specifications

Specifications

Calcium silicate
Reagent
MFCD00015979
OYACROKNLOSFPA-UHFFFAOYSA-N
calcium;dioxido(oxo)silane
26370
Powder
White
10101-39-0
CaO3Si
calcium metasilicate, wollastonite, aedelforsite, casiflux, cab-o-lite, gillebachite, wollastokup, calcium silicon oxide, florite, fuwalip
[O-][Si](=O)[O-].[Ca+2]
116.16
116.16
2.9g/cm3 at 20°C
2kg

Chemical Identifiers

10101-39-0
116.16
OYACROKNLOSFPA-UHFFFAOYSA-N
26370
[O-][Si](=O)[O-].[Ca+2]
CaO3Si
MFCD00015979
calcium metasilicate, wollastonite, aedelforsite, casiflux, cab-o-lite, gillebachite, wollastokup, calcium silicon oxide, florite, fuwalip
calcium;dioxido(oxo)silane
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.

H319-H335 : Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 233-250-6

SDS Documents