Please login to your online account to display your discounted pricing

Calcium silicate, meta, 99% (metals basis), Alfa Aesar

$62.65 - $62.65

Chemical Identifiers

Chemical Name or Material Calcium silicate
Name Note meta
CAS 10101-39-0
Physical Form Powder
Assay Percent Range 99%
Molecular Formula CaSiO3
Formula Weight 116.16
MDL Number MFCD00015979
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3584314

 
alfa aesar
3584314
25g Each for $62.65

AA3584322

 
alfa aesar
3584322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3584336

 
alfa aesar
3584336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.16
Quantity 25g
Color White
Density 2.9g/cm3 at 20°C
Chemical Name or Material Calcium silicate
EINECS Number 233-250-6
Odor Odorless
Name Note meta
Physical Form Powder
Assay Percent Range 99%
Molecular Formula CaSiO3
Appearance White powder
MDL Number MFCD00015979
CAS 10101-39-0
Health Hazard 1 H319-H335 : Causes serious eye irritation. May cause respiratory irritation.
Safety and Handling

H319-H335 : Causes serious eye irritation. May cause respiratory irritation.