missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium shot, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1012706

 View more versions of this product

Catalog No. AA1012706



Specifications

Specifications

Calcium
7440-70-2
99.5% (Metals basis)
MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
calcium
40.078
CHEBI:29320
0 hPa
5g
Silvery-white shot
Ca
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
[Ca]
Odorless
5460341
Shot, Redistilled, 1cm (0.4 in.) & down
Silver-White

Chemical Identifiers

7440-70-2
40.078
OYPRJOBELJOOCE-UHFFFAOYSA-N
5460341
calcium
Ca
MFCD00085314
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
CHEBI:29320
[Ca]
Safety and Handling

Safety and Handling

H261: In contact with water releases flammable gas.

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 213-179-5

Documents