missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium shot, 1cm (0.4 in.) & down, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 1012622

 View more versions of this product

Catalog No. AA1012622



Specifications

Specifications

Calcium shot
7440-70-2
Ca
UN1401
OYPRJOBELJOOCE-UHFFFAOYSA-N
calcium
40.078
CHEBI:29320
1cm (0.4 in.) & down
Undesignated
100g
(metals basis)
MFCD00085314
atom
[Ca]
Odorless
5460341
Shot
99%

Chemical Identifiers

7440-70-2
40.078
OYPRJOBELJOOCE-UHFFFAOYSA-N
5460341
calcium
Ca
MFCD00085314
atom
CHEBI:29320
[Ca]
Safety and Handling

Safety and Handling

GHS H Statement
H261
In contact with water releases flammable gas.

GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM

RTECSNumber : EV8040000

TSCA : TSCA

Documents