Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium shot, 1cm (0.4 in.) & down, 99% (metals basis)

$54.69 - $54.69

Chemical Identifiers

Chemical Name or Material Calcium shot
CAS 7440-70-2
MDL Number MFCD00085314
UN Number UN1401
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1012622

  SDS
alfa aesar™
1012622
100g Each for $54.69

AA1012636

  SDS
alfa aesar™
1012636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1012657

  SDS
alfa aesar™
1012657
4 x 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Particle Size 1cm (0.4 in.) & down
Health Hazard 1 Danger
Odor Odorless
TSCA TSCA
Health Hazard 3 GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H261
In contact with water releases flammable gas.
Assay Percent Notes (metals basis)
Color Metallic
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM
CAS 7440-70-2
RTECS Number EV8040000
Quantity 100g
MDL Number MFCD00085314
Chemical Name or Material Calcium shot
Assay 99%
UN Number UN1401
Physical Form Shot
Safety and Handling

Danger

GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H261
In contact with water releases flammable gas.