Please login to your online account to display your discounted pricing

Calcium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Ca) Solute: Calcium Carbonate Solvent: Dilute Nitric Acid

$82.71 - $114.41

Chemical Identifiers

Chemical Name or Material Calcium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 471-34-1
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00085314
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SC191-500

  SDS
fisher scientific company
SC191500
Poly Bottle 500mL Each for $114.41
Case of 6 Each for $686.46

SC191-100

  SDS
fisher scientific company
FLSC191100
Poly Bottle 100mL Each for $82.71
Case of 6 Each for $496.26
Description & Specifications

Specifications

Purity Grade Certified Grade
Boiling Point 100°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Colorless
Vapor Pressure 14mmHg at 20°C
Viscosity [NULL]
Chemical Name or Material Calcium, Reference Standard Solution
Name Note 1000ppm ±1%
ChemAlert Storage Symbol White
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Concentration 990 to 1010ppm
MDL Number MFCD00085314
CAS 471-34-1
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED CALCIUM REF. STD.
CAS 7732-18-5
CAS 7697-37-2
Safety and Handling