missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium pyruvate, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2520414

 View more versions of this product

Catalog No. AAB2520414



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium pyruvate
52009-14-0
MFCD00037199
calcium pyruvate, calcium 2-oxopropanoate, unii-9v2cqz19n4, 2-oxo-propanoic acid calcium salt, calcium pyruvate, anhydrous, propanoic acid, 2-oxo-, calcium salt, pyruvic acid calcium salt, calcium pyruvic, acmc-20ali9, calcium dipyruvate
CC(=O)C(=O)[O-].CC(=O)C(=O)[O-].[Ca+2]
214.186
214.19
anhydrous
C6H6CaO6
25 g
UZWMCCLZMHPPKW-UHFFFAOYSA-L
calcium;2-oxopropanoate
9859191
97%

Chemical Identifiers

52009-14-0
214.186
UZWMCCLZMHPPKW-UHFFFAOYSA-L
9859191
CC(=O)C(=O)[O-].CC(=O)C(=O)[O-].[Ca+2]
C6H6CaO6
MFCD00037199
calcium pyruvate, calcium 2-oxopropanoate, unii-9v2cqz19n4, 2-oxo-propanoic acid calcium salt, calcium pyruvate, anhydrous, propanoic acid, 2-oxo-, calcium salt, pyruvic acid calcium salt, calcium pyruvic, acmc-20ali9, calcium dipyruvate
calcium;2-oxopropanoate
Safety and Handling

Safety and Handling

EINECSNumber : 257-599-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only