missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Calcium propionate hydrate, 97%
Manufacturer: Alfa Aesar™ A1499436
Specifications
Calcium propionate hydrate | |
500g | |
C6H10CaO4 | |
14,1698 | |
BCZXFFBUYPCTSJ-UHFFFAOYSA-L | |
calcium;propanoate | |
19999 | |
186.22 (Anhydrous) |
>300°C | |
4075-81-4 | |
MFCD00167354 | |
calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907 | |
CCC(=O)[O-].CCC(=O)[O-].[Ca+2] | |
186.22 | |
CHEBI:81716 | |
97% |
Chemical Identifiers
4075-81-4 | |
186.22 | |
BCZXFFBUYPCTSJ-UHFFFAOYSA-L | |
19999 | |
calcium;propanoate |
C6H10CaO4 | |
MFCD00167354 | |
calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907 | |
CHEBI:81716 | |
CCC(=O)[O-].CCC(=O)[O-].[Ca+2] |
Safety and Handling
TSCA : TSCA