Please login to your online account to display your discounted pricing

Calcium Propionate, +98%, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  403900250

 View more versions of this product

Catalog No. AC403900250

  • / Each
 

Description & Specifications

Chemical Identifiers

Arsenic (As) 3ppm max.
Fluoride 10ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Insoluble in Water 0.3% max.
Iron (Fe) 50ppm max.
Lead (Pb) 5ppm max.
Loss on Drying 4% max. (105°C, 2 Hrs)
Packaging Plastic bottle
Color White
pH 6.9 to 9.0 (10% solution)
Quantity 25g
Chemical Name or Material Calcium propionate
CAS 4075-81-4
Physical Form White crystals or crystalline powder
Assay 98.0% min. (on dry substance)
Assay Percent Range 98+%
Molecular Formula C6H10CaO4
Linear Formula (CH3CH2COO)2Ca
Formula Weight 186.22g/mol
MDL Number MFCD00036137
Merck Index 15, 1700
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Calcium propionate
Assay 98.0% min. (on dry substance)
Assay Percent Range 98+%
Molecular Formula C6H10CaO4
Linear Formula (CH3CH2COO)2Ca
Formula Weight 186.22g/mol
MDL Number MFCD00036137
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Harmful if inhaled.

SDS & Safety Documents