Learn More
Calcium, plasma standard solution, Specpure™ Ca 10,000μg/mL, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 014407AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium plasma standard solution | |
Ca 10000μg/mL | |
MFCD00085314 | |
Specpure™ |
CaCO3 in 5% HNO3 | |
100 mL | |
UN3264 | |
Liquid |
Chemical Identifiers
MFCD00085314 |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only