missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13852AD

 View more versions of this product

Catalog No. AA13852AD



Specifications

Specifications

Calcium
Clear liquid
MFCD00085314
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
Liquid
Colorless
471-34-1,7732-18-5,7732-18-5
CCaO3
Nitric acid: 5.0%; Calcium carbonate: 0.25%; Water: 94.75%
C(=O)([O-])[O-].[Ca+2]
100.086
CHEBI:3311
23 hPa (17mm Hg) at 20°C
50mL

Chemical Identifiers

471-34-1
100.086
VTYYLEPIZMXCLO-UHFFFAOYSA-L
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
CCaO3
MFCD00085314
10112
calcium;carbonate
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-714-2

Documents