Please login to your online account to display your discounted pricing

Calcium phosphide, Red-brown lump, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Calcium phosphide
CAS 1305-99-3
Purity Grade Technical
Physical Form Red-brown lump
Molecular Formula Ca3P2
Formula Weight 182.19g/mol
MDL Number MFCD00015986
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1911522

 
alfa aesar
19115-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Sensitivity Moisture sensitive
Color Red-brown
Quantity 100g
Chemical Name or Material Calcium phosphide
CAS 1305-99-3
Purity Grade Technical
Physical Form Red-brown lump
Molecular Formula Ca3P2
Formula Weight 182.19g/mol
MDL Number MFCD00015986
Health Hazard 1 H260-H300-H400: In contact with water releases flammable gases which may ignite spontaneously. Fatal if swallowed. Very toxic to aquatic life.
Health Hazard 2 Do not breathe dust. Wear suitable protective clothing and gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
DOT Information DOT Class: 4.3, Packaging Group: I
EINECS Number 215-142-0