Learn More
Calcium phosphate tribasic, 34-40% Ca, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 036731A3
Description
Utilized in powdered spices as an anticaking agent and baby powder. It is the starting material for the production of phosphoric acid and fertilizers. Employed in tissue replacement for repairing bony defects and act as drug carrier for drug delivery to the bone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium phosphate tribasic | |
White | |
Powder | |
34-40% Ca | |
MFCD00010904 | |
Partially soluble in water. | |
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O | |
310.17 | |
1004.67 |
tribasic | |
12167-74-7 | |
2 kg | |
Ca3O8P2 | |
durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy | |
QORWJWZARLRLPR-UHFFFAOYSA-H | |
tricalcium diphosphate | |
14781 | |
White powder |
Chemical Identifiers
12167-74-7 | |
310.17 | |
QORWJWZARLRLPR-UHFFFAOYSA-H | |
14781 | |
[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
Ca3O8P2 | |
MFCD00010904 | |
durapatite, hydroxylapatite, hydroxyapatite, alveograf, periograf, ossopan, calcium hydroxyapatite, radiesse, monite, apatite, hydroxy | |
tricalcium diphosphate |
Safety and Handling
H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber : 235-330-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only