missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium phosphate (pyro), 96% min

Manufacturer:  Alfa Aesar™ 89836A1

 View more versions of this product

Catalog No. AA89836A1



Specifications

Specifications

Calcium phosphate (pyro)
Ca2O7P2
calcium pyrophosphate, calcium diphosphate, dicalcium diphosphate, unii-x69nu20d19, dicalcium pyrophosphate, alpha-calcium pyrophosphate, calcium phosphate ca2p2o7, diphosphoric acid, calcium salt, calcium pyrophosphate ca2p2o7, diphosphoric acid, calcium salt 1:2
[O-]P(=O)([O-])OP(=O)([O-])[O-].[Ca+2].[Ca+2]
254.097
CHEBI:32598
Powder
3.09g/mL
White
1kg
7790-76-3
MFCD00015983
JUNWLZAGQLJVLR-UHFFFAOYSA-J
dicalcium;phosphonato phosphate
24632
254.1
≥96%
Odorless
1230°C

Chemical Identifiers

7790-76-3
254.097
JUNWLZAGQLJVLR-UHFFFAOYSA-J
24632
dicalcium;phosphonato phosphate
Ca2O7P2
MFCD00015983
calcium pyrophosphate, calcium diphosphate, dicalcium diphosphate, unii-x69nu20d19, dicalcium pyrophosphate, alpha-calcium pyrophosphate, calcium phosphate ca2p2o7, diphosphoric acid, calcium salt, calcium pyrophosphate ca2p2o7, diphosphoric acid, calcium salt 1:2
CHEBI:32598
[O-]P(=O)([O-])OP(=O)([O-])[O-].[Ca+2].[Ca+2]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents