Please login to your online account to display your discounted pricing

Calcium Phosphate, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$618.40 - $618.40

Chemical Identifiers

Chemical Name or Material Calcium Phosphate
CAS 7757-93-9
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula CaHPO4
Formula Weight 136.06
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002697

 
avantor performance
143005
Poly Bottle 2.5kg Each for $618.40
Description & Specifications

Specifications

Formula Weight 136.06
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Physical Form Powder
Molecular Formula CaHPO4
CAS 7757-93-9
Chemical Name or Material Calcium Phosphate
Specific Gravity 1L = 2.30kg
Quantity 2.5kg