Please login to your online account to display your discounted pricing

Calcium Phosphate Monobasic, MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  19380380

 View more versions of this product

Catalog No. ICN19380380

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 234.1
Quantity 100g
Chemical Name or Material Calcium Phosphate Monobasic
Molecular Formula Ca(H2PO4)2
CAS 7758-23-8
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Calcium Phosphate Monobasic
CAS 7758-23-8
Molecular Formula Ca(H2PO4)2
Formula Weight 234.1
Accessories

Room Temperature