Please login to your online account to display your discounted pricing

Calcium Phosphate Monobasic, MP Biomedicals


Quantity 50g

Manufacturer: mp biomedicals  0219380350

 View more versions of this product

Catalog No. ICN19380350

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50g
Chemical Name or Material Calcium Phosphate Monobasic
CAS 7758-23-8
Molecular Formula Ca(H2PO4)2
Formula Weight 234.1g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Calcium Phosphate Monobasic
CAS 7758-23-8
Molecular Formula Ca(H2PO4)2
Formula Weight 234.1g/mol
Accessories

Room Temperature