Please login to your online account to display your discounted pricing

Calcium Phosphate Monobasic, MP Biomedicals

$99.83 - $255.18

Chemical Identifiers

Chemical Name or Material Calcium Phosphate Monobasic
CAS 7758-23-8
Molecular Formula Ca(H2PO4)2
Formula Weight 234.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19380380

 
mp biomedicals
19380380
100g Each for $99.83

ICN19380390

 
mp biomedicals
19380390
500g Each for $255.18
Description & Specifications

Specifications

Formula Weight 234.1
Quantity 100g
Molecular Formula Ca(H2PO4)2
CAS 7758-23-8
Chemical Name or Material Calcium Phosphate Monobasic
Recommended Storage Room Temperature