Please login to your online account to display your discounted pricing

Calcium Phosphate Monobasic, MP Biomedicals


Quantity 500g

Manufacturer: mp biomedicals  19380390

 View more versions of this product

Catalog No. ICN19380390

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 234.1
Quantity 500g
Chemical Name or Material Calcium Phosphate Monobasic
Molecular Formula Ca(H2PO4)2
CAS 7758-23-8
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Calcium Phosphate Monobasic
CAS 7758-23-8
Molecular Formula Ca(H2PO4)2
Formula Weight 234.1
Accessories

Room Temperature