Please login to your online account to display your discounted pricing

Calcium Phosphate Dibasic, MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  0219380480

 View more versions of this product

Catalog No. ICN19380480

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100g
Chemical Name or Material Calcium Phosphate Dibasic
CAS 7757-93-9
Molecular Formula CaHPO4
Formula Weight 136.1g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Calcium Phosphate Dibasic
CAS 7757-93-9
Molecular Formula CaHPO4
Formula Weight 136.1g/mol
Accessories

Room Temperature