Please login to your online account to display your discounted pricing

Calcium Phosphate Dibasic, MP Biomedicals

$36.09 - $133.68

Chemical Identifiers

Chemical Name or Material Calcium Phosphate Dibasic
CAS 7757-93-9
Molecular Formula CaHPO4
Formula Weight 136.1g/mol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19380490

 
mp biomedicals
0219380490
500g Each for $83.56

ICN19380480

 
mp biomedicals
0219380480
100g Each for $36.09

ICN19380491

 
mp biomedicals
0219380491
1kg Each for $133.68
Description & Specifications

Specifications

Quantity 500g
Chemical Name or Material Calcium Phosphate Dibasic
CAS 7757-93-9
Molecular Formula CaHPO4
Formula Weight 136.1g/mol
Recommended Storage Room Temperature
Safety and Handling

Room Temperature