missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 213480050

Catalog No. AC213480050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Calcium phosphate
7758-87-4
35-40%
Ca3(PO4)2
calcium phosphate, synthos, tricalcium phosphate, tricalcium diphosphate, calcigenol simple, calcium orthophosphate, beta-tcp, tribasic calcium phosphate, bonarka, calcium phosphate, tribasic
QORWJWZARLRLPR-UHFFFAOYSA-H
tricalcium;diphosphate
24456
310.18
Analysis
c.p.
5kg
Ca3O8P2
15,1696
Solubility in water: 0.1g/L (25°C)
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Ca+2].[Ca+2].[Ca+2]
310.18
CHEBI:9679
35-40% (Ca)
Plastic Drum
Safety and Handling

Safety and Handling

EINECSNumber : 231-840-8

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only