missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium peroxide, typically 65%, Thermo Scientific Chemicals

Rubber stabilizer

Supplier:  Thermo Scientific Chemicals 02115722

 View more versions of this product

Catalog No. AA2115722



Description

Description

Calcium peroxide is used as a rubber stabilizer, used in different areas of industry and agriculture. In agriculture it is used as an oxygen fertilizer, and is also used in the presowing treatments of rice seed. It is used as flour bleaching agent and improving agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium peroxide
Powder
CaO2
UN1457
calcium peroxide, calcium dioxide, calper, calper g, unii-7fro2eno91, peroxidocalcium, calciumperoxid, kalziumperoxid, calcium peroxide ca o2, hsdb 965
LHJQIRIGXXHNLA-UHFFFAOYSA-N
calcium;peroxide
14779
72.08
Odorless
1305-79-9
100 g
MFCD00015988
14,1689
Slightly soluble in water; Soluble in acids
[O-][O-].[Ca+2]
72.076
CHEBI:48233
65% (typically)
Moisture sensitive

Chemical Identifiers

1305-79-9
72.076
LHJQIRIGXXHNLA-UHFFFAOYSA-N
14779
calcium;peroxide
CaO2
MFCD00015988
calcium peroxide, calcium dioxide, calper, calper g, unii-7fro2eno91, peroxidocalcium, calciumperoxid, kalziumperoxid, calcium peroxide ca o2, hsdb 965
CHEBI:48233
[O-][O-].[Ca+2]
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: CALCIUM PEROXIDE

EINECSNumber : 215-139-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only