missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Calcium Oxide, Reagent grade, Honeywell™

Reagent Grade

$248.28 - $248.28

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.077
MDL Number MFCD00010911
InChI Key ODINCKMPIJJUCX-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, calcium oxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
IUPAC Name oxocalcium
SMILES O=[Ca]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045663 View Documents Honeywell™
248568500G
500g Poly Bottle Each for $248.28
 
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

1305-78-8
56.077
ODINCKMPIJJUCX-UHFFFAOYSA-N
14778
oxocalcium
CaO
MFCD00010911
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, calcium oxide, airlock, calx
CHEBI:31344
O=[Ca]

Specifications

1305-78-8
CaO
UN1910
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, calcium oxide, airlock, calx
O=[Ca]
56.077
CHEBI:31344
Reagent
Poly Bottle
White
500g
CaO
MFCD00010911
Powder
ODINCKMPIJJUCX-UHFFFAOYSA-N
oxocalcium
14778
56.08g/mol
3.3g/mL at 25°C
2850°C
2572°C
Calcium Oxide
Safety and Handling

Safety and Handling

P261-P280-P305 + P351 + P338

H315-H318-H335

DOTInformation : Class - PG 8 - Calcium oxide

EINECSNumber : 215-138-9

RTECSNumber : EW3100000