Please login to your online account to display your discounted pricing

Calcium Oxide, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$147.56 - $465.00

Chemical Identifiers

Chemical Name or Material Calcium Oxide
CAS 1305-78-8
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula CaO
Formula Weight 56.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002536

 
avantor performance
141005
Poly Bottle 2.5kg Each for $465.00

02002537

 
avantor performance
141001
Poly Bottle 500g Each for $147.56
Description & Specifications

Specifications

Chemical Name or Material Calcium Oxide
CAS 1305-78-8
Purity Grade BAKER ANALYZED™ Reagent
Physical Form Powder
Molecular Formula CaO
Formula Weight 56.08
Packaging Poly Bottle
Specific Gravity 1L = 3.37kg
Quantity 2.5kg