Please login to your online account to display your discounted pricing

Calcium oxide, 99.95% (metals basis), Alfa Aesar™

$82.09 - $337.51

Chemical Identifiers

Chemical Name or Material Calcium oxide
CAS 1305-78-8
Physical Form Powder
Assay Percent Range 99.95% (Metal basis)
Molecular Formula CaO
Formula Weight 56.08
MDL Number MFCD00010911
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4477618

  SDS
alfa aesar
4477618
50g Each for $82.09

AA4477630

  SDS
alfa aesar
4477630
250g Each for $337.51

AA44776A1

  SDS
alfa aesar
44776A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 56.08
Quantity 50g
Color White
DOT Information DOT Class: 8, Packaging Group: III
Density 3.25-3.37g/cm3 at 20°C
Physical Form Powder
EINECS Number 215-138-9
Molecular Formula CaO
Assay Percent Range 99.95% (Metal basis)
Chemical Name or Material Calcium oxide
Appearance White powder
MDL Number MFCD00010911
CAS 1305-78-8
Health Hazard 1 H314-H318 : Causes severe skin burns and eye damage. Causes serious eye damage
Safety and Handling

H314-H318 : Causes severe skin burns and eye damage. Causes serious eye damage