missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium oxide, 99.95% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4477630

 View more versions of this product

Catalog No. AA4477630



Specifications

Specifications

Calcium oxide
99.95% (Metal basis)
MFCD00010911
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, calcium oxide, airlock, calx
O=[Ca]
56.077
CHEBI:31344
Powder
250g
1305-78-8
CaO
White powder
ODINCKMPIJJUCX-UHFFFAOYSA-N
oxocalcium
14778
56.08
White

Chemical Identifiers

1305-78-8
56.077
ODINCKMPIJJUCX-UHFFFAOYSA-N
14778
oxocalcium
CaO
MFCD00010911
calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, calcium oxide, airlock, calx
CHEBI:31344
O=[Ca]
Safety and Handling

Safety and Handling

H314-H318 : Causes severe skin burns and eye damage. Causes serious eye damage

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 215-138-9

SDS Documents