missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium oxide, 99.5%, Alfa Aesar

Manufacturer:  Thermo Fisher Scientific 1092330

 View more versions of this product

Catalog No. AA1092330



Specifications

Specifications

Calcium oxide
99.95% (Metal basis)
MFCD00010911
airlock
O=[Ca]
56.077
CHEBI:31344
Powder
White
1305-78-8
CaO
White powder
ODINCKMPIJJUCX-UHFFFAOYSA-N
oxocalcium
14778
56.08
Odorless
250g

Chemical Identifiers

1305-78-8
56.077
ODINCKMPIJJUCX-UHFFFAOYSA-N
14778
oxocalcium
CaO
MFCD00010911
airlock
CHEBI:31344
O=[Ca]
Safety and Handling

Safety and Handling

H314-H318

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 215-138-9

storageNote1 : Air & Moisture Sensitive

Documents