Please login to your online account to display your discounted pricing

Calcium Oxide, ∽97%, MP Biomedicals


Quantity 1kg

Manufacturer: mp biomedicals  0219380291

 View more versions of this product

Catalog No. ICN19380291

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1kg
Chemical Name or Material Calcium Oxide
CAS 1305-78-8
Assay Percent Range ∽97%
Molecular Formula CaO
Formula Weight 56.1g/mol
Synonym (Lime; Quicklime)
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Calcium Oxide
Assay Percent Range ∽97%
Molecular Formula CaO
Formula Weight 56.1g/mol
Synonym (Lime; Quicklime)
Accessories

Room Temperature