Please login to your online account to display your discounted pricing

Calcium Oxide, ∽97%, MP Biomedicals

$180.03 - $180.03

Chemical Identifiers

Chemical Name or Material Calcium Oxide
Assay Percent Range ∽97%
Molecular Formula CaO
Formula Weight 56.1g/mol
Synonym (Lime; Quicklime)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19380291

 
mp biomedicals
0219380291
1kg Each for $180.03
Description & Specifications

Specifications

Quantity 1kg
Chemical Name or Material Calcium Oxide
CAS 1305-78-8
Assay Percent Range ∽97%
Molecular Formula CaO
Formula Weight 56.1g/mol
Synonym (Lime; Quicklime)
Recommended Storage Room Temperature
Safety and Handling

Room Temperature