missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium oxalate monohydrate, 99%, Alfa Aesar

Manufacturer:  Alfa Aesar 3243036*

 View more versions of this product

Catalog No. AA3243036



Specifications

Specifications

500g
2.12
5794-28-5
C2H2CaO5
LQHWSGSWNOHVHO-UHFFFAOYSA-L
calcium;oxalate;hydrate
165350
Calcium oxalate
monohydrate
99%
ethanedioic acid calcium salt 1:1 monohydrate, ethanedioic acidcalcium salt hydrate 1:?:?, calcium oxalate monohydrate 100g, calcium ethanedioate hydrate 1:1:1, calcium oxalate monohydrate, calcium hydrate oxalate, unii-4pp86kk527, acmc-1bmv0
C(=O)(C(=O)[O-])[O-].O.[Ca+2]
146.111
146.11 (128.10anhydrous)

Chemical Identifiers

5794-28-5
146.111
ethanedioic acid calcium salt 1:1 monohydrate, ethanedioic acidcalcium salt hydrate 1:?:?, calcium oxalate monohydrate 100g, calcium ethanedioate hydrate 1:1:1, calcium oxalate monohydrate, calcium hydrate oxalate, unii-4pp86kk527, acmc-1bmv0
calcium;oxalate;hydrate
C2H2CaO5
LQHWSGSWNOHVHO-UHFFFAOYSA-L
165350
C(=O)(C(=O)[O-])[O-].O.[Ca+2]
SDS Documents