Learn More
Calcium oxalate monohydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03243022
Description
Calcium oxalate monohydrate is used as a precursor in the production of oxalic acid, organic oxalates, ceramic glazes, and for rare-earth-metal separation process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium oxalate | |
monohydrate | |
Lump/Powder | |
99% | |
MFCD00150039 | |
Slightly soluble in water. | |
O.[Ca++].[O-]C(=O)C([O-])=O | |
146.11 | |
146.11 (128.10 Anhydrous) | |
2.12 |
200°C | |
5794-28-5 | |
100 g | |
C2H2CaO5 | |
calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:? | |
LQHWSGSWNOHVHO-UHFFFAOYSA-L | |
calcium;oxalate;hydrate | |
165350 | |
Hygroscopic |
Chemical Identifiers
5794-28-5 | |
146.11 | |
LQHWSGSWNOHVHO-UHFFFAOYSA-L | |
165350 | |
O.[Ca++].[O-]C(=O)C([O-])=O |
C2H2CaO5 | |
MFCD00150039 | |
calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:? | |
calcium;oxalate;hydrate |
Safety and Handling
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
EINECSNumber : 209-260-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only