missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 403880025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Calcium oxalate monohydrate | |
5794-28-5 | |
98% | |
(COO)2Ca·H2O | |
2811 | |
calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:? | |
LQHWSGSWNOHVHO-UHFFFAOYSA-L | |
calcium;oxalate;hydrate | |
165350 | |
98% | |
13% max. (200°C) |
98% | |
2.5 kg | |
C2H2CaO5 | |
MFCD00150039 | |
14, 1685 | |
Solubility in water: practically insoluble. Other solubilities: practically insoluble in acetic acid,soluble in dilute hcl or hno3 | |
O.[Ca++].[O-]C(=O)C([O-])=O | |
146.11 | |
146.11 | |
Extra Pure | |
Plastic bottle |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Warning
RUO â Research Use Only