missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium nitride, Brown pieces, 99% (Metals basis), Alfa Aesar™

Chemical Identifiers

CAS 12013-82-0
Physical Form Pieces
Assay Percent Range 99% (Metals basis)
Molecular Formula Ca3N2
Formula Weight 148.25
MDL Number MFCD00015985
Chemical Name or Material Calcium nitride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1312406 SDS Alfa Aesar
1312406
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1312418 SDS Alfa Aesar
1312418
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1312430 SDS Alfa Aesar
1312430
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

12013-82-0
99% (Metals basis)
148.25
Calcium nitride
Pieces
Ca3N2
MFCD00015985

Specifications

Pieces
5g
MFCD00015985
Ca3N2
Ammonia-like
148.25
Brown
12013-82-0
99% (Metals basis)
Calcium nitride

Safety and Handling

H228-H314-H318: Flammable solid. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 234-592-9