missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium nitrate tetrahydrate, MilliporeSigma™

For analysis EMSURE™ ACS

Manufacturer:  MilliporeSigma 1021210500

 View more versions of this product

Catalog No. M1021210500



Specifications

Specifications

Calcium nitrate tetrahydrate
CaH8N2O10
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium;dinitrate;tetrahydrate
16211656
236.15
Odorless
ACS
13477-34-4
UN1454
Soluble in water: 2710g/L at 40°C
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]
236.146
CHEBI:86159
1.82g/cm3 at 20°C
Plastic Bottle
500g

Chemical Identifiers

13477-34-4
236.146
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
CHEBI:86159
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]
CaH8N2O10
ICSSIKVYVJQJND-UHFFFAOYSA-N
16211656
calcium;dinitrate;tetrahydrate
Safety and Handling

Safety and Handling

H319: Causes serious eye irritation.

H272: May intensify fire; oxidiser.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 233-332-1

RTECSNumber : EW3000000

Recommended Storage : Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. Away from combustible substances

Documents
Promotions

Promotions