Learn More
Calcium nitrate tetrahydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Manufacturing of other nitrates, fertilizers, matches
Supplier: Thermo Scientific Chemicals 012364A3
Description
Calcium nitrate tetrahydrate is used as nitrogenous fertilizer and corrosive inhibitor in diesel fuels. It is utilized in plant cell for culture experiments. It is also used in cooling baths and waste water handling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
132°C (decomposition) | |
42°C | |
13477-34-4 | |
2 kg | |
CaH8N2O10 | |
UN1454 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
calcium;dinitrate;tetrahydrate | |
16211656 | |
236.15 (164.09 Anhydrous) | |
ACS | |
Hygroscopic |
Calcium nitrate tetrahydrate | |
tetrahydrate | |
Crystalline | |
99.0-103.0% | |
MFCD00149604 | |
14,1682 | |
Soluble in water,alcohol,acetone and ammonia. | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] | |
236.146 | |
CHEBI:86159 | |
99.0 to 103.0% | |
Odorless | |
1.82 g/mL |
Chemical Identifiers
13477-34-4 | |
236.146 | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
16211656 | |
calcium;dinitrate;tetrahydrate |
CaH8N2O10 | |
MFCD00149604 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
CHEBI:86159 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE
EINECSNumber : 233-332-1
RTECSNumber : EW3000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only