Please login to your online account to display your discounted pricing

Calcium nitrate tetrahydrate, 99%, for analysis, ACROS Organics™


Quantity 5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC217510050

 View more versions of this product

Catalog No. AC217510050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Assay ≥98.5%
Formula Weight 236.15
Quantity 5kg
Color White
pH 4.5 to 7.0 (5% soln. at 20°C)
Density 1.8600g/mL
Purity Grade For analysis
Merck Index 15, 1684
Linear Formula Ca(NO3)2·4H2O
Heavy Metals (as Pb) 5ppm max.
Iron (Fe) 5ppm max.
MDL Number MFCD00149604
Chemical Name or Material Calcium nitrate tetrahydrate
CAS 13477-34-4
Molecular Formula CaN2O6·4H2O
Health Hazard 1 Danger
Trace Analysis 100ppm max.
Packaging Plastic bottle
Chlorine (Cl) 50ppm max.
Sulfate 50ppm max.
Barium (Ba) 50ppm max.
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May intensify fire; oxidizer.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Calcium nitrate tetrahydrate
CAS 13477-34-4
Purity Grade For analysis
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula CaN2O6·4H2O
Linear Formula Ca(NO3)2·4H2O
Formula Weight 236.15
MDL Number MFCD00149604
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May intensify fire; oxidizer.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents