missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium nitrate tetrahydrate, 99.98% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 3048222

 View more versions of this product

Catalog No. AA3048222



Specifications

Specifications

Calcium nitrate tetrahydrate
99.98%
MFCD00149604
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium;dinitrate;tetrahydrate
16211656
236.15
Odorless
100g
13477-34-4
CaH8N2O10
White crystalline or powder
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]
236.146
CHEBI:86159
Crystalline or Powder
White

Chemical Identifiers

13477-34-4
236.146
ICSSIKVYVJQJND-UHFFFAOYSA-N
CHEBI:86159
CaH8N2O10
MFCD00149604
16211656
Safety and Handling

Safety and Handling

H272-H315-H319-H335-H303 : May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 233-332-1

SDS Documents