missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium nitrate tetrahydrate, 99.98% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 3048214

 View more versions of this product

Catalog No. AA3048214



Specifications

Specifications

Calcium nitrate tetrahydrate
99.98%
MFCD00149604
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]
236.146
CHEBI:86159
Crystalline or Powder
Odorless
25g
13477-34-4
CaH8N2O10
White crystalline or powder
ICSSIKVYVJQJND-UHFFFAOYSA-N
calcium;dinitrate;tetrahydrate
16211656
236.15
1.82g/cm3 at 20°C
White

Chemical Identifiers

13477-34-4
236.146
ICSSIKVYVJQJND-UHFFFAOYSA-N
16211656
calcium;dinitrate;tetrahydrate
CaH8N2O10
MFCD00149604
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
CHEBI:86159
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2]
Safety and Handling

Safety and Handling

H272-H315-H319-H335-H303 : May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 233-332-1

SDS Documents