missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium nitrate tetrahydrate, 98%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 447530250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
132.0°C | |
45.0°C | |
5.0 to 9.0 | |
97.5% min. (Complexometry) | |
Ca(NO3)2·4H2O | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
calcium;dinitrate;tetrahydrate | |
16211656 | |
236.15 | |
Extra Pure | |
1.8600g/mL |
Calcium nitrate tetrahydrate | |
13477-34-4 | |
25 kg | |
CaN2O6·4H2O | |
15,1684 | |
Solubility in water: 1470g/L (0°C). Other solubilities: soluble in alcohol and acetone | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] | |
236.15 | |
CHEBI:86159 | |
98% | |
1.86 |
Chemical Identifiers
13477-34-4 | |
236.15 | |
calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate | |
CHEBI:86159 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.[Ca+2] |
CaN2O6·4H2O | |
ICSSIKVYVJQJND-UHFFFAOYSA-N | |
16211656 | |
calcium;dinitrate;tetrahydrate |
Safety and Handling
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
RUO – Research Use Only