missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium nitrate hydrate, Puratronic™, 99.9995% (metals basis)

Manufacturer:  Alfa Aesar™ 4451522

 View more versions of this product

Catalog No. AA4451522



Specifications

Specifications

Calcium nitrate hydrate
(metals basis)
Ca(NO3)2·xH2O
UN1454
acmc-20alid
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Ca+2]
182.101
164.09 (Anhydrous)
99.9995%
1.82 g/mL
White
100g
35054-52-5
CaH2N2O7
MFCD00149604
14,1682
JJIQGEZLLWXYKV-UHFFFAOYSA-N
calcium;dinitrate;hydrate
16211464
Crystalline
Puratronic™
132°C (decomposition)
42°C

Chemical Identifiers

35054-52-5
182.101
JJIQGEZLLWXYKV-UHFFFAOYSA-N
16211464
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.[Ca+2]
CaH2N2O7
MFCD00149604
acmc-20alid
calcium;dinitrate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H302-H315-H319-H335
May intensify fire
oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P370+P378q-P501c

H272-H302-H318

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE

EINECSNumber : 233-332-1

RTECSNumber : EW3000000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents