Learn More
Calcium nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01068314
Description
Calcium nitrate hydrate is used as a nitrate enriching salt. It is also used as a component in fertilizers, waste water treatment to prevent odor and inset accelerating concrete admixtures. Further, it is used for regenerable cold packs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
132°C (decomposition) | |
42°C | |
Crystalline Aggregates | |
(metals basis) | |
MFCD00149604 | |
14,1682 | |
Soluble in water. | |
[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
164.09 | |
164.09 (Anhydrous) | |
Puratronic™ | |
1.82 g/mL |
Calcium nitrate hydrate | |
35054-52-5 | |
25 g | |
CaN2O6 | |
UN1454 | |
calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci | |
ZCCIPPOKBCJFDN-UHFFFAOYSA-N | |
calcium;dinitrate;hydrate | |
16211464 | |
99.995% | |
Hygroscopic |
Chemical Identifiers
35054-52-5 | |
164.09 | |
ZCCIPPOKBCJFDN-UHFFFAOYSA-N | |
16211464 |
CaN2O6 | |
MFCD00149604 | |
calcium nitrate hydrate, calcium hydrate dinitrate, acmc-20alid, calcium nitrate monohydrate, ca.2no3.h2o, nitric acid calcium salt hydrate, calcium nitrate-water 1/2/1, nitric acid, calciumsalt, hydrate 9ci | |
[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
H272-H302-H315-H319-H335
May intensify fire
oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P370+P378q-P501c
H272-H302-H318
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CALCIUM NITRATE
EINECSNumber : 233-332-1
RTECSNumber : EW3000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only