Please login to your online account to display your discounted pricing

Calcium Nitrate ACS AR Crystal, Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Poly

Manufacturer: avantor performance  423606

 View more versions of this product

Catalog No. MK4236212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Grade
Formula Weight 236.15
Quantity 2.5kg
Chemical Name or Material Calcium Nitrate
Name Note 4-Hydrate
Packaging Poly
Physical Form Crystalline
Molecular Formula Ca(NO3)2·4H2O
CAS 13477-34-4

Specifications

Chemical Name or Material Calcium Nitrate
Name Note 4-Hydrate
CAS 13477-34-4
Purity Grade ACS Grade
Physical Form Crystalline
Molecular Formula Ca(NO3)2·4H2O
Formula Weight 236.15