missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium Nitrate ACS AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 13477-34-4
Physical Form Crystalline
Molecular Formula Ca(NO3)2·4H2O
Formula Weight 236.15
Chemical Name or Material Calcium Nitrate, 4-Hydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK4236212 Avantor Performance
423606
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK4236500 Avantor Performance
423604
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

13477-34-4
Ca(NO3)2·4H2O
Calcium Nitrate, 4-Hydrate
Crystalline
236.15
ACS Grade

Specifications

13477-34-4
Ca(NO3)2·4H2O
Poly
Calcium Nitrate, 4-Hydrate
Crystalline
236.15
2.5kg
ACS Grade