Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium molybdenum oxide, Puratronic™, 99.9965% (metals basis)

Chemical Identifiers

Chemical Name or Material Calcium molybdenum oxide
CAS 7789-82-4
Purity Grade Puratronic™
Formula Weight 200.01
MDL Number MFCD00015976
Merck Index 14,1681
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1068206

  SDS
alfa aesar™
1068206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1068214

  SDS
alfa aesar™
1068214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 200.01
Assay Percent Notes (metals basis)
Physical Form Powder
Density 4.38g/mL to 4.53g/mL
MDL Number MFCD00015976
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Merck Index 14,1681
Chemical Name or Material Calcium molybdenum oxide
Purity Grade Puratronic™
Assay 99.9965%
CAS 7789-82-4
TSCA TSCA
Melting Point 965°C
RTECS Number EW2975000
Odor Odorless
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.