missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium molybdenum oxide, 99%

Manufacturer:  Alfa Aesar™ 4368622

 View more versions of this product

Catalog No. AA4368622



Specifications

Specifications

Calcium molybdenum oxide
CaMoO4
14,1681
BIOOACNPATUQFW-UHFFFAOYSA-N
calcium;dioxido(dioxo)molybdenum
24620
Powder
4.45g/mL
White
100g
7789-82-4
MFCD00015976
calcium 2+ molybdate
[O-][Mo](=O)(=O)[O-].[Ca+2]
200.024
200.01
99%
Odorless
965°C

Chemical Identifiers

7789-82-4
200.024
BIOOACNPATUQFW-UHFFFAOYSA-N
24620
[O-][Mo](=O)(=O)[O-].[Ca+2]
CaMoO4
MFCD00015976
calcium 2+ molybdate
calcium;dioxido(dioxo)molybdenum
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : EW2975000

TSCA : TSCA

Documents