missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium metaborate, 99.992% (metals basis)

Manufacturer:  Alfa Aesar™ 1298806

 View more versions of this product

Catalog No. AA1298806



Specifications

Specifications

Calcium metaborate
(metals basis)
Ca(BO2)2
calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
B(=O)[O-].B(=O)[O-].[Ca+2]
125.694
125.7
99.992%
White
13701-64-9
B2CaO4
MFCD00049397
ILOKQJWLMPPMQU-UHFFFAOYSA-N
calcium;oxido(oxo)borane
10290800
Powder
Odorless
5g

Chemical Identifiers

13701-64-9
125.694
ILOKQJWLMPPMQU-UHFFFAOYSA-N
10290800
B(=O)[O-].B(=O)[O-].[Ca+2]
B2CaO4
MFCD00049397
calcium metaborate, 2bo2.ca, calcium dioxoborinate, dimetaboric acid calcium salt, calcium 2+ diboron oxide hydroxide
calcium;oxido(oxo)borane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents